Learn More
Clear Line DES 70%, Solveco
Product Code. 30281153
Change view
Click to view available options
Quantity:
5 L
Unit Size:
15 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30281153 5 L 15 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30281153 Supplier Solveco Chemicals Supplier No. 1055
 Restricted Item
Estimated Shipment: 04-11-2026
Clear Line DES 70 is a surface disinfectant designed for smooth and visibly clean surfaces such as tables, countertops, and sinks. Use Clear Line DES 70 in areas where the spread of microbiological contamination needs to be prevented. The product is effective against bacteria, fungi, and various types of viruses.
Clear Line DES 70 is a surface disinfectant designed for smooth and visibly clean surfaces such as tables, countertops, and sinks. Use Clear Line DES 70 in areas where the spread of microbiological contamination needs to be prevented. The product is effective against bacteria, fungi, and various types of viruses.
Clear Line DES 70 has been tested according to and meets the European standards for disinfectants: EN 1040, EN 1275, EN 1276, EN 14348, and EN 1650.
Specifications
Color-coded Yes
Form Clear Liquid
Scent Unscented
Sterility Non-sterile
Quantity 5 L