Learn More
Metro™ Clear Label Holders for Wire Shelving
Product Code. 15946771
Change view
Click to view available options
Label Size (English):
3 x 1.25 in.
13 x 1.25 in.
19 x 1.25 in.
25 x 1.25 in.
31 x 1.25 in.
43 x 1.25 in.
55 x 1.25 in.
Label Size (Metric):
109.22 x 3.2 cm
139.5 x 3.2 cm
33 x 3.2 cm
48 x 3.2 cm
63.5 x 3.2 cm
7.5 x 3.2 cm
72.5 x 3.2 cm
Length (English) Shelf:
18 in.
24 in.
30 in.
36 in.
48 in.
60 in.
Length (Metric) Shelf:
45.7 cm
61 cm
76 cm
91.4 cm
121.9 cm
152 cm
Unit Size:
Each
Product Code. Label Size (English) Label Size (Metric) Length (English) Shelf Length (Metric) Shelf unitSize
15946771 25 x 1.25 in. 63.5 x 3.2 cm 30 in. 76 cm Each
15906771 3 x 1.25 in. 7.5 x 3.2 cm - - Each
15916771 13 x 1.25 in. 33 x 3.2 cm 18 in. 45.7 cm Each
15926771 19 x 1.25 in. 48 x 3.2 cm 24 in. 61 cm Each
15936771 31 x 1.25 in. 72.5 x 3.2 cm 36 in. 91.4 cm Each
15956771 43 x 1.25 in. 109.22 x 3.2 cm 48 in. 121.9 cm Each
15966771 55 x 1.25 in. 139.5 x 3.2 cm 60 in. 152 cm Each
7 options
Product Code. 15946771
Brand: Metro™ 9990CL30
Only null left
Add to basket
Add to basket
Snap-on plastic holders provide easy identification of shelf contents.
  • Clear plastic allows label colors to show through
  • Hold most commercial 1.25 in. (32 mm) labels
  • Labels not included
Specifications
Product Type Label Holder
Color Clear
For Use With (Equipment) Metro™ Super Erecta™, Super Adjustable™ 2, qwikSLOT™, Drop Mat™ wire shelving, cantilever shelves
Label Size (English) 25 x 1.25 in.
Label Size (Metric) 63.5 x 3.2 cm
Length (English) Shelf 30 in.
Length (Metric) Shelf 76 cm
Temperature Range (English) -18 to 120.2°F
Temperature Range (Metric) -28 to 49°C
Material Plastic