Learn More
Karl Hecht™ Clear Glass Wide Neck Bottles
Product Code. 10227571
Change view
Click to view available options
Capacity (Metric):
50 mL
250 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10227571 50 mL Each
10563842 250 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10227571 Supplier Karl Hecht™ Supplier No. 41311006
  • ONLINE EXCLUSIVE PRICE
    110.00 NOK
    67.00 NOK / Pack of 1
    Save 43.00 NOK
    39% Off
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
Manufactured from clear glass for excellent visibility of contents.
  • With ground-in glass stopper
  • Wide neck for ease of access to contents
  • Available in different capacities
Specifications
Capacity (Metric) 50 mL
Color Clear
Includes Ground In Glass Stopper
Material Glass
Clarity Clear
Closure Included Yes
Mouth Wide
Shape Round
Height (Metric) 76 mm
Product Type Storage Jar
Closure Material Glass
Diameter (Metric) 41.5 mm
Closure Type Ground-in Stopper