Learn More
Thermo Scientific™ Clear Flat-Bottom Immuno Nonsterile 96-Well Plates
GREENER_CHOICE
Product Code. 10190825
Change view
Click to view available options
Surface Treatment:
Enhanced Binding
Immulon 1 B
Immulon 2 HB
Immulon 4 HBX
MaxiSorp
MediSorp
MultiSorp
PolySorp
Universal Binding
Certified:
No
Yes
Volume (Metric) Working:
230 μL
280 μL
330 μL
350 μL
Barcode:
Includes Barcode
No Barcode
Unit Size:
Pack of 180
Pack of 50
Pack of 60
Product Code. Surface Treatment Certified Volume (Metric) Working Barcode unitSize
10190825 Universal Binding No 330 μL No Barcode Pack of 50
10547915 Immulon 1 B No 230 μL No Barcode Pack of 50
10795026 Immulon 2 HB No 280 μL No Barcode Pack of 50
10468075 Immulon 4 HBX No 280 μL No Barcode Pack of 50
10394751 MaxiSorp Yes 350 μL No Barcode Pack of 60
10547781 MaxiSorp No 350 μL No Barcode Pack of 60
10498802 PolySorp No 350 μL No Barcode Pack of 180
10294401 MaxiSorp Yes 350 μL No Barcode Pack of 180
10264451 MaxiSorp Yes 350 μL Includes Barcode Pack of 180
10275411 MediSorp Yes 350 μL No Barcode Pack of 60
10565131 MultiSorp No 350 μL No Barcode Pack of 60
10449672 PolySorp No 350 μL No Barcode Pack of 60
10130665 Enhanced Binding Yes 330 μL No Barcode Pack of 50
13 options
Each lot is functionally tested for IgG binding.
Product Code. 10190825
Brand: Thermo Scientific™ 9502227
Only null left
Add to basket
Add to basket
Each lot is functionally tested for IgG binding.
Choose from four binding surfaces of these 96-well plates for reliable and reproducible results in colormetric assays.
Choose from four binding surfaces for reliable and reproducible results in colormetric assays with Thermo Scientific™ Clear Flat-Bottom Immuno Nonsterile 96-Well Plates, which feature Thermo Scientific™ Nunc™, Immulon™ and Microtiter™ clear plates that are ideal for quantitative and qualitative solid phase immunoassays.
96-Well Solid Plates:
  • Clear polystyrene
  • Choice of four surfaces for optimal binding
  • Alphanumerically labeled rows and columns
Surfaces:
  • Thermo Scientific PolySorp, Immulon 1B, and Universal Binding (UB) are for adsorption of hydrophobic molecules.
  • Thermo Scientific Medisorp and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules.
  • Thermo Scientific MaxiSorp, Immulon 4 HBX, and Enhanced Binding(EB) are hydrophilic and ideal for antibody sandwich assays.
  • Thermo Scientific MultiSorp is very hydrophilic and works well for glycans and water soluble proteins.
Ordering Information:
Please note that availability may vary by country. 467320, 456537, 460984 and 439454 include certificate
TRUSTED_SUSTAINABILITY
Specifications
Barcode No Barcode
Certified No
Format Standard
Detection Method Colorimetric
Array 8 x 12
Packaging 50 Cases
Binding Type Hydrophobic
Color Clear
Product Type Microplate
Material Polystyrene
Volume (Metric) Well 450 μL
No. of Wells 96
Volume (Metric) Working 330 μL
Sterility Non-sterile
Surface Treatment Universal Binding
Well Shape Flat Bottom (F-well)
No. per Case 50
No. per Pack 25
Plate Blocking None
Target Molecule Lipids