Learn More
BRAND™ Clear Dropping Bottles with Pipette
Soda-lime glass construction. The interchangeable ground joint stopper has an integrated dropping pipette with rubber cap.
169.63 NOK - 192.44 NOK
Specifications
Color | Clear |
---|---|
Material | Soda Lime Glass |
Closure Included | Yes |
Closure Material | Rubber |
Closure Type | Ground Joint Stopper with Rubber Cap |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
10213541
|
BRAND™
124633 |
50 mL |
216.00 NOK 169.63 NOK / Each Save 46.37 NOK 21% Off |
Estimated Shipment: 10-10-2025
Log in to see stock. |
||||||
10364991
|
BRAND™
124639 |
100 mL |
245.00 NOK 192.44 NOK / Each Save 52.56 NOK 21% Off |
Estimated Shipment: 20-10-2025
Log in to see stock. |
||||||
Description
- Clear soda-lime glass construction
- Includes dropping pipette and rubber cap
Specifications
Clear | |
Yes | |
Ground Joint Stopper with Rubber Cap |
Soda Lime Glass | |
Rubber |
Product Title
BRAND™ Clear Dropping Bottles with Pipette