Learn More
CleanSpace™ Technology CleanSpace™ HALO Neck Support
Plastic neck support is for use with the CleanSpace HALO PAPR units.
875.00 NOK
Specifications
Disposable | Reusable |
---|---|
Material | ABS Plastic |
Product Type | Neck Support |
For Use With (Equipment) | CS3000 PAPR System |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15950042
|
CleanSpace™ Technology
CS3009 |
Small |
875.00 NOK
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
15960042
|
CleanSpace™ Technology
CS3010 |
Large |
875.00 NOK
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
Description
- Used with CleanSpace respirators: light weight, no hoses or belts
- Designed for comfort over long periods
- Allows sweating and breathability
- Easy to wash and quick drying
- Designed for long wear in harsh environments
- Easy and quick replacement
Specifications
Reusable | |
ABS Plastic | |
Neck Support | |
CS3000 PAPR System |