Learn More
PURUS™ Cleanroom Sticky Notes
Product Code. 16450051 Shop All Purus Products
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
16450051 White Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16450051 Supplier PURUS™ Supplier No. LCIB 1000C
  • ONLINE EXCLUSIVE PRICE
    6036.48 NOK
    4578.25 NOK / Pack of 100
    Save 1458.23 NOK
    24% Off
Estimated Shipment: 18-01-2027
Cleanroom Sticky Notes are ideal for writing, small note taking and reminders in cleanroom environments.
  • Excellent for writing and note taking in a cleanroom environment
  • 100 sheets per pad, individually wrapped
  • 100 pads per case
  • Recyclable
  • Suitable for use in ISO 4 to 5 (Class 10 to Class 1000) cleanrooms
Specifications
Material Synthetic
Ruled No
Color White
Description Cleanroom Sticky Notes
Dimensions (L x W) 76 x 76 mm
Autoclavable Autoclavable
For Use With (Application) For use in Class 10 to Class 1000 Cleanrooms. For writing or note taking in a cleanroom environment.
Plain paper Yes
Product Type Cleanroom Sticky Note
Quantity 100 Sheets/Pad, 100 Pads/Cs.
Weight (Metric) 68 gsm