Learn More
Cole-Parmer Cleanroom Male Slip Luer to Barb Fittings, Straight Adapter, White Nylon
Product Code. 30053419 Shop All Cole-Parmer Products
Change view
Click to view available options
Diameter (English) Inner:
1/16 in.
1/4 in.
1/8 in.
3/16 in.
3/32 in.
5/32 in.
Unit Size:
Pack of 10
Product Code. Diameter (English) Inner unitSize
30053419 3/32 in. Pack of 10
30054349 5/32 in. Pack of 10
30053742 3/16 in. Pack of 10
30053339 1/4 in. Pack of 10
30054165 1/16 in. Pack of 10
30053275 1/8 in. Pack of 10
6 options
Product Code. 30053419
Brand: Cole-Parmer 5011531
Only null left
Add to basket
Add to basket
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Connect to a female luer body to form a leak-free fluid path. Single barb design tightly grips the tubing. Meets FDA regulations 21 CFR 178.3297 and RoHS compliant
Specifications
Certifications/Compliance FDA;Non-DEHP;Non-Phthalate;RoHS
Diameter (English) Inner 3/32 in.
Material Nylon
Packaging Quantity 10/Pk.
Inlet Male Slip Luer, Hose Barb
Color White