Learn More
Cole-Parmer Cleanroom Male Slip Luer to Barb Fittings, Straight Adapter, White Nylon
Shop All Cole-Parmer ProductsDescription
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Connect to a female luer body to form a leak-free fluid path. Single barb design tightly grips the tubing. Meets FDA regulations 21 CFR 178.3297 and RoHS compliant
Specifications
Specifications
| Certifications/Compliance | FDA;Non-DEHP;Non-Phthalate;RoHS |
| Diameter (English) Inner | 3/32 in. |
| Material | Nylon |
| Packaging Quantity | 10/Pk. |
| Inlet | Male Slip Luer, Hose Barb |
| Color | White |