Description
Seal luer ports with a secure and leak-proof connection. Luer fittings allow for a simple yet secure twist-lock connection. Ideal for capping off fluid transfer system. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application
Specifications
Specifications
| Certifications/Compliance | Non-DEHP;Non-Phthalate;USP |
| Material | Kynar |
| Inlet | Male Luer Lock, Plug |
| Color | Natural |
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.