Learn More
Cole-Parmer Cleanroom Male Luer Locking Plugs, Straight Adapter
Product Code. 30053816 Shop All Cole-Parmer Products
Change view
Click to view available options
Material:
CrystalVu (CVU)
Kynar
Nylon
Polypropylene
Color:
Clear
Natural
White
Unit Size:
Pack of 10
Product Code. Material Color unitSize
30053816 Kynar Natural Pack of 10
30052941 Nylon White Pack of 10
30054044 CrystalVu (CVU) Clear Pack of 10
30053282 Polypropylene Clear Pack of 10
30054047 Nylon Natural Pack of 10
5 options
Product Code. 30053816
Brand: Cole-Parmer 5011051
Only null left
Add to basket
Add to basket
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
Seal luer ports with a secure and leak-proof connection. Luer fittings allow for a simple yet secure twist-lock connection. Ideal for capping off fluid transfer system. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application
Specifications
Certifications/Compliance Non-DEHP;Non-Phthalate;USP
Material Kynar
Inlet Male Luer Lock, Plug
Color Natural
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.