Learn More
Cole-Parmer Cleanroom Hose Barb Fittings, Straight Reducer, Animal-Derivative-Free Polypropylene
Product Code. 30054262 Shop All Cole-Parmer Products
Change view
Click to view available options
Tubing Size:
1 to 1/2 in.
1 to 3/4 in.
1 to 5/8 in.
1/2 to 1/4 in.
1/2 to 3/8 in.
1/2 to 5/16 in.
1/4 to 1/16 in.
1/4 to 1/8 in.
1/4 to 3/16 in.
1/4 to 3/32 in.
1/4 to 5/32 in.
1/8 to 1/16 in.
1/8 to 3/32 in.
3/16 to 1/16 in.
3/16 to 1/8 in.
3/16 to 3/32 in.
3/32 to 1/16 in.
3/4 to 1/2 in.
3/4 to 3/8 in.
3/8 to 1/4 in.
3/8 to 1/8 in.
3/8 to 5/16 in.
3/8 to 5/32 in.
5/16 to 1/4 in.
5/16 to 1/8 in.
5/16 to 3/16 in.
5/16 to 5/32 in.
5/32 to 1/8 in.
5/32 to 3/32 in.
5/8 to 1/2 in.
5/8 to 1/4 in.
5/8 to 3/8 in.
5/8 to 5/16 in.
Unit Size:
Pack of 10
Product Code. Tubing Size unitSize
30054262 1/4 to 1/16 in. Pack of 10
30053068 3/8 to 5/32 in. Pack of 10
30053879 5/8 to 5/16 in. Pack of 10
30053002 3/16 to 3/32 in. Pack of 10
30052960 1/8 to 1/16 in. Pack of 10
30053647 1/2 to 3/8 in. Pack of 10
30054326 5/16 to 1/4 in. Pack of 10
30053127 5/8 to 1/4 in. Pack of 10
30054356 1/4 to 5/32 in. Pack of 10
30053714 3/8 to 1/4 in. Pack of 10
30053889 5/16 to 3/16 in. Pack of 10
30053808 5/8 to 3/8 in. Pack of 10
30052950 1/2 to 5/16 in. Pack of 10
30053607 1 to 1/2 in. Pack of 10
30053453 1/2 to 1/4 in. Pack of 10
30054370 1/4 to 1/8 in. Pack of 10
30053844 3/8 to 5/16 in. Pack of 10
30054192 1/8 to 3/32 in. Pack of 10
30053782 1 to 3/4 in. Pack of 10
30053468 3/32 to 1/16 in. Pack of 10
30054125 5/8 to 1/2 in. Pack of 10
30053709 5/32 to 3/32 in. Pack of 10
30054132 1/4 to 3/32 in. Pack of 10
30052935 1/4 to 3/16 in. Pack of 10
30053301 1 to 5/8 in. Pack of 10
30053431 3/4 to 1/2 in. Pack of 10
30053733 3/4 to 3/8 in. Pack of 10
30054112 5/16 to 5/32 in. Pack of 10
30053870 5/16 to 1/8 in. Pack of 10
30053146 3/16 to 1/16 in. Pack of 10
30053484 3/8 to 1/8 in. Pack of 10
30053023 3/16 to 1/8 in. Pack of 10
30053569 5/32 to 1/8 in. Pack of 10
33 options
Product Code. 30054262
Brand: Cole-Parmer 5010892
Only null left
Add to basket
Add to basket
Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Quick and easy assembly without tools. Single barb design tightly grips the tubing. Meets FDA requirements 21 CFR 177.1520(a)(3)(i) and (c)3.1a and RoHS compliant
Specifications
Certifications/Compliance ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS
Min. Operating Temperature (Metric) -23°C
Packaging Quantity 10/Pk.
Tubing Size 1/4 to 1/16 in.
Outlet Port Hose Barb Reducer
Sterilization Method 121°C
Shape Straight
Color Natural