Learn More
- Home
- Products
- Tubing
- Tubing Connectors and Fittings
- Cole-Parmer Cleanroom Hose Barb Fittings, Straight Reducer, Animal-Derivative-Free Polypropylene
Cole-Parmer Cleanroom Hose Barb Fittings, Straight Reducer, Animal-Derivative-Free Polypropylene
Product Code. 30053879 Shop All Cole-Parmer Products
Change view
Click to view available options
Tubing Size:
1 to 1/2 in.
1 to 3/4 in.
1 to 5/8 in.
1/2 to 1/4 in.
1/2 to 3/8 in.
1/2 to 5/16 in.
1/4 to 1/16 in.
1/4 to 1/8 in.
1/4 to 3/16 in.
1/4 to 3/32 in.
1/4 to 5/32 in.
1/8 to 1/16 in.
1/8 to 3/32 in.
3/16 to 1/16 in.
3/16 to 1/8 in.
3/16 to 3/32 in.
3/32 to 1/16 in.
3/4 to 1/2 in.
3/4 to 3/8 in.
3/8 to 1/4 in.
3/8 to 1/8 in.
3/8 to 5/16 in.
3/8 to 5/32 in.
5/16 to 1/4 in.
5/16 to 1/8 in.
5/16 to 3/16 in.
5/16 to 5/32 in.
5/32 to 1/8 in.
5/32 to 3/32 in.
5/8 to 1/2 in.
5/8 to 1/4 in.
5/8 to 3/8 in.
5/8 to 5/16 in.
Unit Size:
Pack of 10
| Product Code. | Tubing Size | unitSize |
|---|---|---|
| 30053879 | 5/8 to 5/16 in. | Pack of 10 |
| 30053068 | 3/8 to 5/32 in. | Pack of 10 |
| 30053002 | 3/16 to 3/32 in. | Pack of 10 |
| 30052960 | 1/8 to 1/16 in. | Pack of 10 |
| 30053647 | 1/2 to 3/8 in. | Pack of 10 |
| 30054326 | 5/16 to 1/4 in. | Pack of 10 |
| 30053127 | 5/8 to 1/4 in. | Pack of 10 |
| 30054356 | 1/4 to 5/32 in. | Pack of 10 |
| 30053714 | 3/8 to 1/4 in. | Pack of 10 |
| 30053889 | 5/16 to 3/16 in. | Pack of 10 |
| 30053808 | 5/8 to 3/8 in. | Pack of 10 |
| 30052950 | 1/2 to 5/16 in. | Pack of 10 |
| 30053607 | 1 to 1/2 in. | Pack of 10 |
| 30053453 | 1/2 to 1/4 in. | Pack of 10 |
| 30054370 | 1/4 to 1/8 in. | Pack of 10 |
| 30053844 | 3/8 to 5/16 in. | Pack of 10 |
| 30054192 | 1/8 to 3/32 in. | Pack of 10 |
| 30053782 | 1 to 3/4 in. | Pack of 10 |
| 30053468 | 3/32 to 1/16 in. | Pack of 10 |
| 30054125 | 5/8 to 1/2 in. | Pack of 10 |
| 30053709 | 5/32 to 3/32 in. | Pack of 10 |
| 30054132 | 1/4 to 3/32 in. | Pack of 10 |
| 30052935 | 1/4 to 3/16 in. | Pack of 10 |
| 30053301 | 1 to 5/8 in. | Pack of 10 |
| 30053431 | 3/4 to 1/2 in. | Pack of 10 |
| 30053733 | 3/4 to 3/8 in. | Pack of 10 |
| 30054112 | 5/16 to 5/32 in. | Pack of 10 |
| 30053870 | 5/16 to 1/8 in. | Pack of 10 |
| 30053146 | 3/16 to 1/16 in. | Pack of 10 |
| 30053484 | 3/8 to 1/8 in. | Pack of 10 |
| 30053023 | 3/16 to 1/8 in. | Pack of 10 |
| 30053569 | 5/32 to 1/8 in. | Pack of 10 |
| 30054262 | 1/4 to 1/16 in. | Pack of 10 |
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Quick and easy assembly without tools. Single barb design tightly grips the tubing. Meets FDA requirements 21 CFR 177.1520(a)(3)(i) and (c)3.1a and RoHS compliant
Specifications
Specifications
| Certifications/Compliance | ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS |
| Min. Operating Temperature (Metric) | -135°C |
| Packaging Quantity | 10/Pk. |
| Tubing Size | 5/8 to 5/16 in. |
| Outlet Port | Hose Barb Reducer |
| Sterilization Method | 121°C |
| Shape | Straight |
| Color | Natural |