Learn More
- Home
- Products
- Tubing
- Tubing Connectors and Fittings
- Cole-Parmer Cleanroom Hose Barb Fittings, Straight Reducer, Animal-Derivative-Free Polypropylene
Cole-Parmer Cleanroom Hose Barb Fittings, Straight Reducer, Animal-Derivative-Free Polypropylene
Product Code. 30053301 Shop All Cole-Parmer Products
Change view
Click to view available options
Tubing Size:
1 to 1/2 in.
1 to 3/4 in.
1 to 5/8 in.
1/2 to 1/4 in.
1/2 to 3/8 in.
1/2 to 5/16 in.
1/4 to 1/16 in.
1/4 to 1/8 in.
1/4 to 3/16 in.
1/4 to 3/32 in.
1/4 to 5/32 in.
1/8 to 1/16 in.
1/8 to 3/32 in.
3/16 to 1/16 in.
3/16 to 1/8 in.
3/16 to 3/32 in.
3/32 to 1/16 in.
3/4 to 1/2 in.
3/4 to 3/8 in.
3/8 to 1/4 in.
3/8 to 1/8 in.
3/8 to 5/16 in.
3/8 to 5/32 in.
5/16 to 1/4 in.
5/16 to 1/8 in.
5/16 to 3/16 in.
5/16 to 5/32 in.
5/32 to 1/8 in.
5/32 to 3/32 in.
5/8 to 1/2 in.
5/8 to 1/4 in.
5/8 to 3/8 in.
5/8 to 5/16 in.
Conditionnement:
Pack of 10
| Code produit. | Tubing Size | unitSize |
|---|---|---|
| 30053301 | 1 to 5/8 in. | Pack of 10 |
| 30053068 | 3/8 to 5/32 in. | Pack of 10 |
| 30053879 | 5/8 to 5/16 in. | Pack of 10 |
| 30053002 | 3/16 to 3/32 in. | Pack of 10 |
| 30052960 | 1/8 to 1/16 in. | Pack of 10 |
| 30053647 | 1/2 to 3/8 in. | Pack of 10 |
| 30054326 | 5/16 to 1/4 in. | Pack of 10 |
| 30053127 | 5/8 to 1/4 in. | Pack of 10 |
| 30054356 | 1/4 to 5/32 in. | Pack of 10 |
| 30053714 | 3/8 to 1/4 in. | Pack of 10 |
| 30053889 | 5/16 to 3/16 in. | Pack of 10 |
| 30053808 | 5/8 to 3/8 in. | Pack of 10 |
| 30052950 | 1/2 to 5/16 in. | Pack of 10 |
| 30053607 | 1 to 1/2 in. | Pack of 10 |
| 30053453 | 1/2 to 1/4 in. | Pack of 10 |
| 30054370 | 1/4 to 1/8 in. | Pack of 10 |
| 30053844 | 3/8 to 5/16 in. | Pack of 10 |
| 30054192 | 1/8 to 3/32 in. | Pack of 10 |
| 30053782 | 1 to 3/4 in. | Pack of 10 |
| 30053468 | 3/32 to 1/16 in. | Pack of 10 |
| 30054125 | 5/8 to 1/2 in. | Pack of 10 |
| 30053709 | 5/32 to 3/32 in. | Pack of 10 |
| 30054132 | 1/4 to 3/32 in. | Pack of 10 |
| 30052935 | 1/4 to 3/16 in. | Pack of 10 |
| 30053431 | 3/4 to 1/2 in. | Pack of 10 |
| 30053733 | 3/4 to 3/8 in. | Pack of 10 |
| 30054112 | 5/16 to 5/32 in. | Pack of 10 |
| 30053870 | 5/16 to 1/8 in. | Pack of 10 |
| 30053146 | 3/16 to 1/16 in. | Pack of 10 |
| 30053484 | 3/8 to 1/8 in. | Pack of 10 |
| 30053023 | 3/16 to 1/8 in. | Pack of 10 |
| 30053569 | 5/32 to 1/8 in. | Pack of 10 |
| 30054262 | 1/4 to 1/16 in. | Pack of 10 |
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application. PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries. Quick and easy assembly without tools. Single barb design tightly grips the tubing. Meets FDA requirements 21 CFR 177.1520(a)(3)(i) and (c)3.1a and RoHS compliant
Spécification
Spécification
| Certifications/Compliance | ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS |
| Min. Operating Temperature (Metric) | -23°C |
| Packaging Quantity | 10/Pk. |
| Tubing Size | 1 to 5/8 in. |
| Outlet Port | Hose Barb Reducer |
| Sterilization Method | 121°C |
| Shape | Straight |
| Color | Natural |
Nom du produit
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?