Description
- Tests available for HDPE, nylon, PP, PTFE and PVC
- Two sizes available for each configuration, either 50 x 50mm or 100 x 100mm
Specifications
Specifications
| Product Type | Cleaning Coupon |
| Dimensions (L x W) | 50 x 50 mm |
| For Use With (Application) | Testing and validating cleaning regime |
| Material | Polypropylene |