Learn More
Sampling Systems™ Cleaning Coupons for Metal
Product Code. 15899551 Shop All Sampling Systems Products
Change view
Click to view available options
Material:
Aluminum
Stainless Steel (304)
Stainless Steel (316)
Dimensions (L x W):
100 x 100 mm
50 x 50 mm
50 x 50 mm
Finish:
Bright Polish
Satin finish
Unit Size:
Each
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Dimensions (L x W) Finish Unit Size
15899551 Stainless Steel (304) 50 x 50 mm Bright Polish Each
15889541 Aluminum 50 x 50 mm Bright Polish Each
15809561 Stainless Steel (304) 50 x 50 mm Satin finish Each
15839561 Stainless Steel (316) 50 x 50 mm Bright Polish Each
15849561 Stainless Steel (316) 50 x 50 mm Satin finish Each
15879541 Aluminum 100 x 100 mm Bright Polish Each
15879551 Stainless Steel (304) 100 x 100 mm Bright Polish Each
15889551 Stainless Steel (304) 100 x 100 mm Satin finish Each
15819561 Stainless Steel (316) 100 x 100 mm Bright Polish Each
15829561 Stainless Steel (316) 100 x 100 mm Satin finish Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 15899551 Supplier Sampling Systems™ Supplier No. 8644S450B
  • ONLINE EXCLUSIVE PRICE
    1710.00 NOK
    1572.20 NOK / Pack of 1
    Save 137.80 NOK
    8% Off
Estimated Shipment: 30-12-2026
Test and validate your cleaning regime with this range of cleaning validation coupons for various metals.
  • Tests available for aluminum, 304 stainless steel and 316 stainless steel
  • All metals also available for either bright polish or satin finish
  • Two sizes available for each configuration, either 50 x 50mm or 100 x 100mm
Specifications
Product Type Cleaning Coupon
Dimensions (L x W) 50 x 50 mm
Finish Bright Polish
For Use With (Application) Testing and validating cleaning regime
Material Stainless Steel (304)