Learn More
Sampling Systems™ Cleaning Coupons for Metal
Product Code. 15889541 Shop All Sampling Systems Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Material:
Aluminum
Stainless Steel (304)
Stainless Steel (316)
Dimensions (L x W):
100 x 100 mm
50 x 50 mm
50 x 50 mm
Finish:
Bright Polish
Satin finish
Conditionnement:
Each
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Material Dimensions (L x W) Finish Conditionnement
15889541 Aluminum 50 x 50 mm Bright Polish Each
15899551 Stainless Steel (304) 50 x 50 mm Bright Polish Each
15809561 Stainless Steel (304) 50 x 50 mm Satin finish Each
15839561 Stainless Steel (316) 50 x 50 mm Bright Polish Each
15849561 Stainless Steel (316) 50 x 50 mm Satin finish Each
15879541 Aluminum 100 x 100 mm Bright Polish Each
15879551 Stainless Steel (304) 100 x 100 mm Bright Polish Each
15889551 Stainless Steel (304) 100 x 100 mm Satin finish Each
15819561 Stainless Steel (316) 100 x 100 mm Bright Polish Each
15829561 Stainless Steel (316) 100 x 100 mm Satin finish Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 15889541 Fournisseur Sampling Systems™ Code fournisseur 8644AL50
  • ONLINE EXCLUSIVE PRICE
    890.00 NOK
    795.45 NOK / Pack of 1
    Save 94.55 NOK
    10% Off
Estimated Shipment: 22-01-2027
Ajouter au panier
Ajouter au panier
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Test and validate your cleaning regime with this range of cleaning validation coupons for various metals.
  • Tests available for aluminum, 304 stainless steel and 316 stainless steel
  • All metals also available for either bright polish or satin finish
  • Two sizes available for each configuration, either 50 x 50mm or 100 x 100mm
Specifications
Product Type Cleaning Coupon
Dimensions (L x W) 50 x 50 mm
Finish Bright Polish
For Use With (Application) Testing and validating cleaning regime
Material Aluminum