Learn More
Sampling Systems™ Cleaning Coupons for Metal
Shop All Sampling Systems Products
Click to view available options
Material:
Aluminum
Stainless Steel (304)
Stainless Steel (316)
Dimensions (L x W):
100 x 100 mm
50 x 50 mm
50 x 50 mm
Finish:
Bright Polish
Satin finish
Unit Size:
Each
Description
- Tests available for aluminum, 304 stainless steel and 316 stainless steel
- All metals also available for either bright polish or satin finish
- Two sizes available for each configuration, either 50 x 50mm or 100 x 100mm
Specifications
Specifications
| Product Type | Cleaning Coupon |
| Dimensions (L x W) | 100 x 100 mm |
| Finish | Bright Polish |
| For Use With (Application) | Testing and validating cleaning regime |
| Material | Stainless Steel (316) |