Learn More
PURUS™ CleanImage Premier Cleanroom Paper
Product Code. p-4367708 Shop All Purus Products
Change view
Click to view available options
Color:
Blue
White
Dimensions (L x W):
8.3 x 11.7 in.
Page Format:
Holeless
Weight (Metric):
105 gsm
80 gsm
Unit Size:
Pack of 10
Product Code. Color Dimensions (L x W) Page Format Weight (Metric) unitSize
15793589 White 8.3 x 11.7 in. Holeless 80 gsm Pack of 10
15703589 Blue 8.3 x 11.7 in. Holeless 80 gsm Pack of 10
15723589 Blue 8.3 x 11.7 in. Holeless 105 gsm Pack of 10
3 options
Product Code. 15793589
Brand: PURUS™ PCIW 1081C
Only null left
Add to basket
Add to basket
For writing, printing and photocopying in cleanroom environments, PURUS™ Cleanroom Paper provides high printability and heat toner adhesion, and is also recyclable.
  • Propriatary Binding Technology
  • Suitable for use in ISO 4 to 6 (Class 10 to Class 1000) Cleanroom
  • Produced in ISO 4 (Class 10 Cleanroom)
  • Recyclable
  • High printability and heat toner adhesion
Compliance
Suitable for use in ISO 4 to 6 (Class 10 to Class 1000).
Specifications
Material Polyethylene
Ruled No
Width (English) Page 11.7 in.
Length (English) Page 8.3 in.
Color White
Cover Color White
Dimensions (L x W) 8.3 x 11.7 in.
Autoclavable Autoclavable
For Use With (Application) Writing, Printing and Photocopying in Cleanroom Environments
Page Format Holeless
Plain paper Yes
Product Type Cleanroom Paper
Quantity 250 Sheets/Pk., 10 Pk./Cs.
Size A4
Weight (English) 22 lb.
Weight (Metric) 80 gsm