Learn More
Scotch-Brite™ Clean & Shine Floor Pad
Product Code. 17158770
Change view
Click to view available options
Diameter (Metric):
280 mm
330 mm
355 mm
380 mm
406 mm
432 mm
460 mm
480 mm
505 mm
530 mm
Unit Size:
Pack of 5
Product Code. Diameter (Metric) unitSize
17158770 355 mm Pack of 5
17148770 280 mm Pack of 5
17168770 330 mm Pack of 5
17188760 380 mm Pack of 5
17138770 406 mm Pack of 5
17118770 432 mm Pack of 5
17128770 460 mm Pack of 5
17198760 480 mm Pack of 5
17108770 505 mm Pack of 5
17178770 530 mm Pack of 5
10 options
Product Code. 17158770
Brand: Scotch-Brite™ 7100148038
Only null left
Add to basket
Lägg i varukorgen
Denna artikel kan inte returneras. Se returpolicy
This 2 in 1 daily cleaning pad for low speed scrubbers gradually increases shine with repeated use
  • Cleans and shines in the same step, using only a low-speed scrubber
  • Save costs and labor by reducing or eliminating the need to burnish
  • Removes black marks 3 times faster than traditional cleaning pads
  • Two-sided pad lasts longer than traditional cleaning pads
  • Effective on most coated and uncoated hard floors including VCT, vinyl, rubber, marble, stone, terrazzo, granite, and concrete with only water or neutral cleaner
TRUSTED_SUSTAINABILITY
Specifikationer
Product Type Cleaning Pads
Color Brown
Diameter (Metric) 355 mm
For Use With (Application) Cleaning and Polishing Floors
For Use With (Equipment) Concrete, Granite, Industrial Floors, Linoleum, Marble, Stoneware, Terrazzo, VCT, and Vinyl Flooring
Style Round
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.