Learn More
Scotch-Brite™ Clean & Shine Floor Pad
Description
- Cleans and shines in the same step, using only a low-speed scrubber
- Save costs and labor by reducing or eliminating the need to burnish
- Removes black marks 3 times faster than traditional cleaning pads
- Two-sided pad lasts longer than traditional cleaning pads
- Effective on most coated and uncoated hard floors including VCT, vinyl, rubber, marble, stone, terrazzo, granite, and concrete with only water or neutral cleaner
Specifications
Specifications
| Product Type | Cleaning Pads |
| Color | Brown |
| Diameter (Metric) | 505 mm |
| For Use With (Application) | Cleaning and Polishing Floors |
| For Use With (Equipment) | Concrete, Granite, Industrial Floors, Linoleum, Marble, Stoneware, Terrazzo, VCT, and Vinyl Flooring |
| Style | Round |