Learn More
PURUS™ Critical Print Cleanroom Notebooks
Product Code. 15763609 Shop All Purus Products
Click to view available options
Color:
Blue
White
Size:
A4
A5
A6
Ruled:
Grid
Yes
Quantity:
10/Cs.
20/Cs.
40/Cs.
Unit Size:
Pack of 10
Pack of 20
Pack of 40
Product Code. 15763609
Brand: PURUS™ PNBCPA52220 WG
Only null left
Add to basket
Add to basket
Critical Print Notebooks are ideal for writing and data recording in cleanroom environments.
For writing in cleanroom environments
  • Cleanroom processed and packaged
  • Suitable for use in ISO 4 to 6 (Class 10 to Class 1000) cleanrooms
  • Proprietary Binding Technology without organic fillers
  • Recyclable
  • College Rule
  • Amide and Silicone Free
Specifications
Material Polyethylene
Ruled Grid
Color White
Cover Material Poly
Dimensions (L x W) 210 x 148 mm
Width (Metric) 148 mm
Autoclavable Autoclavable
Certifications/Compliance Suitable for use in ISO 4 to 6 (Class 10 to Class 1000)
For Use With (Application) For writing in cleanroom environments
Product Type Cleanroom Notebook
Quantity 20/Cs.
Size A5
Weight (Metric) 80 gsm