Learn More
Emma Safety Footwear Clay S3 Shoe
Product Code. 17582626
Change view
Click to view available options
Size:
39
40
41
42
45
46
47
48
Unit Size:
1 pair
Product Code. Size unitSize
17582626 39 1 pair
17592626 40 1 pair
17502636 41 1 pair
17512636 42 1 pair
17522636 45 1 pair
17532636 46 1 pair
17542636 47 1 pair
17552636 48 1 pair
8 options
Product Code. 17582626
Brand: Emma Safety Footwear MM98054939
Only null left
Add to basket
Add to basket
If trainers are what you prefer to wear but you need a safer shoe for work, look no further than the Clay.
  • S3-Safetyshoe with sole plate for wet work
  • Metal Free
  • The streetborn safety shoe. If trainers are what you prefer to wear but you need a safer shoe for work, look no further than the Clay. A no compromise metal free safety shoe that looks and feels like a classic sneaker.
  • It gets its name from the highly supportive reinforced heel.
  • This heel together with the shoe's reinforced, shock absorbing sole keeps ankles, shins and joints free from stress and injury.
  • DenimX laces with a story. Made from recycled jeans for our circular safety shoes.
  • Industries:
    • Logistics & transport
    • Electronics
    • Construction & technology
    • Manufacturing
    • Nature & environment
Specifications
Certifications/Compliance ISO 20345:2011
Closure Type Laces
Color Black
For Use With (Application) Logistics and Transport, Electronics, Construction and Technology, Manufacturing, Nature and Environment
Liner Material Hydro-Tec Antibacterial
Material Nubuck, Oiled Nubuck
Size 39
Fit Standard Width
Ankle Style Low-cut
Product Type Safety Shoe