missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claudin 23 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25296-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
CLDN23 is a four-transmembrane protein with WWCC motif, defined by W-X(17-22)-W-X(2)-C-X(8-10)-C. It is a candidate tumor suppressor gene implicated in intestinal-type gastric cancer.Specifications
Claudin 23 | |
Polyclonal | |
Unconjugated | |
CLDN23 | |
claudin 23, CLDN23, CLDNL, hCG1646163 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
137075 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96B33 | |
CLDN23 | |
Claudin 23 Fusion Protein Ag18189 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |