missing translation for 'onlineSavingsMsg'
Learn More

Claudin 18 Mouse anti-Human, Mouse, Clone: 5G7F2, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66167-1-IG-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16847795

  • 3828.53 NOK / 150µL
Estimated Shipment: 13-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Claudin-18 is a tight junction protein expressed as lung-and stomach-specific isoforms which are generated by alternative splicing of the claudin-18 gene. The lung-specific form is a downstream target gene regulated by the T/EBP/NKX2.1 transcription factor. A splice variant lacking the C-terminal cytoplasmic domain also exists in mouse, but has not been confirmed in human. Human claudin-18 demonstrates 88% amino acid sequence identity to the mouse protein. Immunohistochemical studies have demonstrated complete membrane localization of claudin-18 in lung and stomach epithelial cells, while electron microscopy has shown that it is concentrated in the cell-cell borders of these cells. These features suggest a potentially important role for claudin-18 in the structure and function of tight junctions in the lung and stomach. Claudin-18 expression has also been reported in the inner ear.
Specifications

Specifications

Claudin 18
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P56856, P56857
CLDN18
Claudin 18 Fusion Protein Ag15517
150 μL
Primary
Human, Mouse
Antibody
IgG2a
Immunohistochemistry (Paraffin), Immunoprecipitation
5G7F2
Unconjugated
CLDN18
claudin 18, CLDN18, SFTA5, SFTPJ
Mouse
Protein A
RUO
51208, 56492
-20°C
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers