Description
- With a locking rod to secure the cryoboxes
- These racks are for half sized boxes with 5 x 5 cell divider
- Max. footprint of 83 x 83 mm
Specifications
Specifications
| Holds | Boxes |
| Material | Stainless Steel |
| Positions | 11 |
| For Use With (Equipment) | Cyrogenic Freezers |
| Dimensions (L x W x H) | 83 x 83 x 610 mm |
| Height (Metric) | 50 mm (Box) |