Learn More
Kolmi Hopen™ Classic Elasticated Gown with Collar and Pocket
Shop All Medicom Products
Click to view available options
Size:
2X-Small
3X-Small
Unit Size:
Pack of 50
Description
- Made of breathable polypropylene
- With collar and elasticated cuffs
- Individual packaging for controlled hygiene and distribution
Specifications
Specifications
| Size | 2X-Small |
| Certifications/Compliance (Europe) | ISO 9001 |
| Gender | Unisex |
| No. of Pockets | 1 |
| Product Type | General Purpose Gown |
| Material | Polypropylene |
| Color | White |
| Closure Type | Press Studs |
| Disposable | Single-use |
| Weight (Metric) | 40 gsm |
| Show More |