Learn More
Zwiebel™ Class M1 Weight Set
Click to view available options
Mass:
1 to 200 g
1 to 2000 g
1 to 50 g
1 to 500 g
1 to 500 mg
Unit Size:
Each
Description
- Class M2 stainless steel calibration weight
- Delivered with a calibration certificate guaranteeing traceability
- In compliance with OIML R111
- Supplied individually or as a set, in a plastic case or a wooden box
Specifications
Specifications
| Mass | 1 to 500 g |
| For Use With (Application) | For use with Weighing Instruments of Medium Accuracy Class III and Ordinary Accuracy Class IIII |