Learn More
Zwiebel™ Class F2 Box Weights
Product Code. 11820912
Click to view available options
Mass:
1 g
2 g
10 g
100 g
200 g
500 g
1000 g
2000 g
Tolerance:
±0.3 mg
±0.4 mg
±0.6 mg
±1.6 mg
±15 mg
±3 mg
±30 mg
±7.5 mg
Unit Size:
Each
Product Code. 11820912
Brand: Zwiebel™ PBI56827
Only null left
Specifications
Mass 2 g
Tolerance ±0.4 mg
For Use With (Application) For use with Weighing Instruments of Special Accuracy Class I and High Accuracy Class II