Learn More
Zwiebel™ Class F1 Weight Set
Click to view available options
Mass:
1 to 50 g
1 to 500 mg
Unit Size:
Each
Specifications
Specifications
| Mass | 1 to 50 g |
| For Use With (Application) | For use with Weighing Instruments of Special Accuracy Class I and High Accuracy Class II |