Learn More
Zwiebel™ Class F1 Weight Box with Certificate
Product Code. p-8006533
Change view
Click to view available options
Mass:
100 g
500 g
2000 g
Tolerance:
±10 mg
±2.50 mg
±5 mg
Unit Size:
Each
Product Code. Mass Tolerance unitSize
11870892 100 g ±5 mg Each
11880902 2000 g ±10 mg Each
11810892 500 g ±2.50 mg Each
3 options
Product Code. 11870892
Brand: Zwiebel™ PBI56819
Only null left
Specifications
Mass 100 g
Tolerance ±5 mg
For Use With (Application) For use with Weighing Instruments of Special Accuracy Class I and High Accuracy Class II