Learn More
Zwiebel™ Class F1 Weight Box with Certificate
Product Code. 11880902
Click to view available options
Mass:
100 g
500 g
2000 g
Tolerance:
±10 mg
±2.50 mg
±5 mg
Unit Size:
Each
Product Code. 11880902
Brand: Zwiebel™ PBI56822
Only null left
Specifications
Mass 2000 g
Tolerance ±10 mg
For Use With (Application) For use with Weighing Instruments of Special Accuracy Class I and High Accuracy Class II