Learn More
Zwiebel™ Class F1 Box Weights
Product Code. 11870902
Change view
Click to view available options
Mass:
50 mg
200 mg
500 mg
2 g
50 g
100 g
200 g
1000 g
2000 g
Tolerance:
±0.04 mg
±0.06 mg
±0.08 mg
±0.12 mg
±0.30 mg
±0.50 mg
±1.00 mg
±10 mg
±5.00 mg
Unit Size:
Each
Product Code. Mass Tolerance unitSize
11870902 2000 g ±10 mg Each
12011643 200 g ±1.00 mg Each
12031643 1000 g ±5.00 mg Each
11730703 100 g ±0.50 mg Each
12021633 200 mg ±0.06 mg Each
12091633 50 g ±0.30 mg Each
12031633 500 mg ±0.08 mg Each
12051633 2 g ±0.12 mg Each
12001633 50 mg ±0.04 mg Each
9 options
Product Code. 11870902
Brand: Zwiebel™ PBI56821
Only null left
Add to basket
Add to basket
Use with weighing instruments of medium accuracy class II. Zwiebel™ Class F1 Box Weights are ideal for the verification or calibration of class F2 weights.
  • In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box
Specifications
Mass 2000 g
Tolerance ±10 mg
For Use With (Application) For use with Weighing Instruments of Special Accuracy Class I and High Accuracy Class II