Learn More
Zwiebel™ Class E2 Box Weight with COFRAC Calibration Certificate
Product Code. 11795183
Click to view available options
Mass:
1 g
1 to 100 g
10 g
100 g
2 g
20 g
200 g
5 g
5 kg
50 g
Unit Size:
Each
Product Code. 11795183
Brand: Zwiebel™ PBI23876
Only null left
Add to basket
Add to basket
Calibrate weights of lower accuracy classes and are used with class I weighing instruments. Zwiebel™ Class E2 Box Weights are intended for use with class F1 weights.
  • Class E2 stainless steel calibration weight
  • Delivered with a calibration certificate guaranteeing traceability
  • In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box
Specifications
Mass 20 g
Tolerance ±0.08 mg
For Use With (Application) For use with Weighing Instruments of Special Accuracy Class I