Learn More
Kartell™ Low Form Graduated Beakers Class B
Click to view available options
Capacity (Metric):
100 mL
500 mL
Unit Size:
Pack of 12
Description
- Crystal clear
- Good chemical resistance
- Permanent graduations moulded into beaker
- Can withstand temperatures up to 170°C for short periods
- Suitable for applications with food
Specifications
Specifications
| Color | Crystal Clear |
| Diameter (Metric) Outer | 88mm |
| Capacity (Metric) | 500 mL |
| Material | PMP |
| For Use With (Application) | Suitable for Foodstuff |
| Height (Metric) | 119mm |
| Quantity | 12Pack |
| Type | Beaker |
| Autoclavable | Autoclavable |
| Graduated | Yes |
| Show More |