Learn More
Kartell™ PMP Class B Low Form Beakers
Manufactured from PMP for exceptional clarity and good chemical resistance. Kartell™ PMP Class B Low Form Beakers are autoclavable and suitable for foodstuffs.
Brand:  Kartell™ 0154100
Product Code. 10122281
  • 754.00 NOK / Pack of 20
Only null left
Estimated Shipment: 06-02-2026
Add to basket
Description
Description
  • Crystal clear PMP translates to safety and economy with reduced breakage in the lab
  • Withstands 170°C for short periods, 150°C for continuous work
  • Permanent molded graduations provide reproducibility and clarity
  • Autoclavable
  • Class B, Low form
  • Complies with ISO 7056-1981 (E) and BS 5404 Part 1
  • Suitable for foodstuff
*Not suitable for use with stirrer hotplates