Learn More
Kartell™ PMP Class B Low Form Beakers
Manufactured from PMP for exceptional clarity and good chemical resistance. Kartell™ PMP Class B Low Form Beakers are autoclavable and suitable for foodstuffs.
Brand: Kartell™ 0154100
This item is not returnable.
View return policy
Description
- Crystal clear PMP translates to safety and economy with reduced breakage in the lab
- Withstands 170°C for short periods, 150°C for continuous work
- Permanent molded graduations provide reproducibility and clarity
- Autoclavable
- Class B, Low form
- Complies with ISO 7056-1981 (E) and BS 5404 Part 1
- Suitable for foodstuff