Learn More
BRAND™ Class B Polypropylene Low Form Measuring Cylinders
Shop All Brand Products
Click to view available options
Unit Size:
Pack of 5
Description
- Low form graduated cylinder with spout
- High clarity polypropylene construction
- Calibrated to contain (TC, In)
- Embossed scale
- Exposure to temperatures up to 80°C will not cause permanently exceeded error limits
Specifications
Specifications
| Class | B |
| Graduations | 10 mL |
| Autoclavable | Autoclavable |
| Limit of Error | ±10mL |
| Max. Temperature | 80°C |
| Shape | Round |
| Base Type | Hexagonal |
| Type | Graduated Measuring Cylinder |