Learn More
Hirschmann™ Class B Measuring Cylinders, Low Form, Amber Stain Graduation
Product Code. 11736692
Change view
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Each
Pack of 2
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11736692 250 mL Pack of 2
11716682 10 mL Pack of 2
11716692 50 mL Pack of 2
11726692 100 mL Pack of 2
11706692 25 mL Pack of 2
11746692 500 mL Pack of 2
10344361 1000 mL Each
10242951 2000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 missing translation for 'options'
Product Code. 11736692 missing translation for 'mfr' Hirschmann™ missing translation for 'supplierNo' 2400186
  • ONLINE EXCLUSIVE PRICE
    612.00 NOK
    538.14 NOK / Pack of 2
    Save 73.86 NOK
    12% Off
Estimated Shipment: 08-12-2026
Excellent for precise liquid measurement and fluid transfer in the laboratory. Hirschmann Instruments™ Class B Measuring Cylinders, Low Form are manufactured from durable DURAN™ glass tubing.
  • With spout, short line
  • Amber stain graduation
  • Acc. to DIN EN ISO 4788
  • Corrosion-free and resistant to thermal shock
Specifications
Material Glass
Capacity (Metric) 250 mL
Graduations 5 mL
Limit of Error 2
Base Type Hexagonal
Scale Color Brown
Shape Round
Class Class B
Height (Metric) 220 mm
Certifications/Compliance DIN EN ISO 4788
Tolerance ±2 mL
Product Type Graduated Measuring Cylinder