Learn More
Hirschmann™ Class B Measuring Cylinders, Blue Stain Graduation
Product Code. 11748021
Change view
Click to view available options
Capacity (Metric):
5 mL
10 mL
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Each
Pack of 2
Product Code. Capacity (Metric) unitSize
11748021 25 mL Pack of 2
11738021 10 mL Pack of 2
11758021 50 mL Pack of 2
11812861 50 mL Pack of 2
11768021 100 mL Pack of 2
11778021 250 mL Pack of 2
11788021 500 mL Pack of 2
10573661 1000 mL Each
10668051 2000 mL Each
11796482 5 mL Pack of 2
10 options
Product Code. 11748021
Brand: Hirschmann™ 2230170
Only null left
Add to basket
Add to basket
Excellent for precise liquid measurement and fluid transfer in the laboratory. Hirschmann Instruments™ Class B Measuring Cylinders, Blue Stain Graduation are manufactured from durable DURAN™ glass tubing.
  • With spout, short line
  • Blue stain graduation
  • Acc. to DIN EN ISO 4788
  • Corrosion-free and resistant to thermal shock
Specifications
Certifications/Compliance DIN EN ISO 4788
Class B
Height (Metric) 170 mm
Material Glass
Capacity (Metric) 25 mL
Graduations 0.5 mL
Limit of Error 0.375