Learn More
Hirschmann™ Class B Graduated Polypropylene Cylinders with Raised Blue Graduations
Description
- Acc. to DIN 12681/ISO 6706
- PP
- Raised scale
- Main point ring graduation
- Hexagonal base
Specifications
Specifications
| Material | Polypropylene |
| Certifications/Compliance | DIN 12681, ISO 6706 |
| No. per Pack | 12/Pack |
| Description | Graduated cylinder, PP |
| Class | B |
| Disposable | Reusable |
| Diameter (Metric) | 15 mm |
| Graduations | 0.2 mL |
| Shape | Round |
| For Use With (Application) | Precise Liquid Measurement and Fluid Transfer in the Laboratory |
| Show More |