Learn More
Hirschmann™ Class B Graduated Polypropylene Cylinders with Raised Blue Graduations
Product Code. 13148029
Change view
Click to view available options
Unit Size:
Pack of 12
Pack of 3
Pack of 6
Capacity (Metric):
25 mL
1000 mL
Product Code. unitSize Capacity (Metric)
13148029 Pack of 12 -
13128029 Pack of 12 -
13138029 Pack of 12 25 mL
13168029 Pack of 6 -
13188029 Pack of 6 1000 mL
13198029 Pack of 3 -
6 options
Product Code. 13148029
Brand: Hirschmann™ 7500175
Only null left
Add to basket
Add to basket
Excellent for precise liquid measurement and fluid transfer in the laboratory.
  • Acc. to DIN 12681/ISO 6706
  • PP
  • Raised scale
  • Main point ring graduation
  • Hexagonal base
Specifications
Class B
Disposable Reusable
Shape Round
For Use With (Application) Precise Liquid Measurement and Fluid Transfer in the Laboratory
Scale Color Blue
Base Type Hexagonal
Height (Metric) 200 mm
Type Graduated Measuring Cylinder