Learn More
Hecht Karl™ Class B DURAN Borosilicate Glass Tall Form Measuring Cylinders
Product Code. p-8003647
Click to view available options
Capacity (Metric):
2000 mL
Unit Size:
Each
Product Code. 10003921
Brand: Hecht Karl™ 42170072
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Tolerance ±15 mL
Material Borosilicate Glass
Scale Color Blue
Certifications/Compliance DIN EN ISO 4788
Class B
Capacity (Metric) 2000 mL
Scale Blue Graduations
Tolerance (Metric) ±15mL
Product Type Graduated Measuring Cylinder
Limit of Error ±15mL