Learn More
Fisherbrand™ Class B Glass Tall Form Measuring Cylinders
Description
- Graduated
- Stable hexagonal foot
- Corrosion-free and resistant to thermal shock
Specifications
Specifications
| Material | Borosilicate Glass |
| Capacity (Metric) | 2000 mL |
| Graduations | 20 mL |
| Autoclavable | Autoclavable |
| Limit of Error | ±20mL |
| Class | Class B |
| Height (Metric) | 560 mm |
| Certifications/Compliance | ISO 4788 |
| Tolerance | ±20 mL |
| Product Type | Graduated Measuring Cylinder |
| Show More |