Learn More
Fisherbrand™ Class B Glass Tall Form Measuring Cylinders
Product Code. 11837501
Change view
Click to view available options
Capacity (Metric):
5 mL
10 mL
25 mL
50 mL
500 mL
1000 mL
2000 mL
Graduations:
0.1 mL
0.2 mL
0.5 mL
1 mL
5 mL
10 mL
20 mL
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Graduations Unit Size
11837501 500 mL 5 mL Each
11847501 1000 mL 10 mL Each
11887491 10 mL 0.2 mL Each
11857501 2000 mL 20 mL Each
11897491 25 mL 0.5 mL Each
11807501 50 mL 1 mL Each
11877491 5 mL 0.1 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 11837501 Supplier Fisherbrand™ Supplier No. DBG507
  • ONLINE EXCLUSIVE PRICE
    170.00 NOK
    154.76 NOK / Pack of 1
    Save 15.24 NOK
    8% Off
Estimated Shipment: 26-10-2026
Measure and pour liquids with precision with these cylinders. Fisherbrand™ Class B Glass Tall Form Measuring Cylinders are manufactured from calibrated, clear borosilicate glass.
  • Graduated
  • Stable hexagonal foot
  • Corrosion-free and resistant to thermal shock
Specifications
Material Borosilicate Glass
Capacity (Metric) 500 mL
Graduations 5 mL
Autoclavable Autoclavable
Limit of Error ±5mL
Class Class B
Height (Metric) 380 mm
Certifications/Compliance ISO 4788
Tolerance ±5 mL
Product Type Graduated Measuring Cylinder
Diameter (Metric) Outer 53 mm