Learn More
Hirschmann™ Class AS Volumetric Pipettes with Amber Stain Graduation
Description
- One mark
- Acc. to DIN EN ISO 648
- Conformity certified, with dated batch identification
Specifications
Specifications
| Class | AS |
| Material | AR-GLAS |
| Graduated | Yes |
| Graduation Color | Brown |
| Quantity | 12/Pk. |
| Tolerance | 0.015 mL |
| Volume (Metric) | 7 mL |
| Length (Metric) | 410 mm |
| Type | Volumetric Pipet |