Learn More
Hecht Karl™ Class A Soda Glass Tall Form Measuring Cylinders
Click to view available options
Capacity (Metric):
100 mL
2000 mL
Type:
Graduated Measuring Cylinder
Unit Size:
Each
Specifications
Specifications
| Item Description | Measuring Cylinder |
| Scale Color | Amber |
| Type | Graduated Measuring Cylinder |
| Material | Soda Lime Glass |
| Certifications/Compliance | DIN EN ISO 4788 |
| Class | B |
| Capacity (Metric) | 100 mL |
| Scale | Short Line Fused Amber Graduation |
| Graduations | 2 mL |
| Subdivision | 2mL |