Learn More
VITLAB™ Class A PMP Volumetric Flask
Description
- Safely protect samples from the effects of light, while maintaining high transparency to enable volumes to be set accurately
- With ring mark individually calibrated to ‘In’
- Class A tolerances according to DIN EN ISO 1042
- With printed lot number and batch certificate
- Thermal stress up to 121°C (autoclaving) does not permanently exceed the tolerance limit
- To preserve markings, cleaning at no higher than 60°C is recommended
- Also available with DAkkS calibration certificate or individual quality certificate
Specifications
Specifications
| Calibration | Yes |
| Certifications/Compliance | DIN EN ISO 1042 |
| Color | Crystal clear |
| Description | Class A PMP Volumetric Flask with NS 19/26 stopper |
| Capacity (Metric) | 250 mL |
| Material | PMP |
| Height (Metric) | 235 mm |
| Tolerance | ±0.15 mL |
| Class | A |
| Closure Type | Standard Ground Stopper |