Learn More
VITLAB™ Class A PMP Tall-Form Measuring Cylinder
Product Code. p-8000799
Click to view available options
Capacity (Metric):
100 mL
500 mL
1000 mL
Graduations:
1 mL
2 mL
5 mL
10 mL
Unit Size:
Each
Pack of 2
Product Code. 11358543
Brand: VITLAB™ 64904
Only null left
Add to basket
Edge
Add to basket
Manufactured with crystal clear PMP for optimum visibility. VITLab™ Class A PMP Tall-Form Measuring Cylinders are designed with a hexagonal base with bottom studs to provide high stability.
  • Raised scale and ring marks at the primary scale points, calibrated ‘In’
  • Lot certificate supplied bears batch number and actual nominal value ascertained under test conditions
  • The resulting deviations from the nominal value fall well under the allowed tolerances of Class A according to DIN 12681 and ISO 6706
  • DE-M marked
  • Laser engraved batch number and year of manufacture
  • Thermal stress up to 121 °C (autoclaving) does not cause tolerance limits to be permanently exceeded
Specifications
Capacity (Metric) 100 mL
Material PMP
Tolerance ±0.5 mL
Certifications/Compliance DIN 12681, ISO 6706
No. per Pack 2
Class A
Scale Raised
Diameter (Metric) 33 mm
Quantity 2Pack
Graduations 1 mL
Autoclavable Autoclavable
Limit of Error ±0.5mL
Shape Round
Base Type Hexagonal
Height (Metric) 250 mm
Type Graduated Cylinder