Learn More
Hirschmann™ Class A PMP Measuring Cylinder with Elevated Scale
Product Code. 10648591
Click to view available options
Capacity (Metric):
100 mL
Unit Size:
Pack of 2
Product Code. 10648591
Brand: Hirschmann™ 7520180
Only null left
Add to basket
Add to basket
For volumetric accuracy in liquid handling applications.
  • With elevated scale and ring mark at the main points
  • Tall version
  • Clear
  • Hexagonal socket
  • Acc. to DIN 12 681/ISO 6706
  • Manufactured from PMP
  • White graduation marks
  • Certified conformity
Specifications
Capacity (Metric) 100 mL
Material PMP
Tolerance ±0.5 mL
Certifications/Compliance DIN 12681, ISO 6706
Class A
Diameter (Metric) 33 mm
Includes Elevated scale
Graduations 1 mL
Subdivision 1 mL
Shape Round
For Use With (Application) Precise Liquid Measurement and Fluid Transfer in the Laboratory
Scale Color White
Base Type Hexagonal
Height (Metric) 250 mm
Type Graduated Measuring Cylinder