Learn More
Hirschmann™ Class A Graduated PMP Cylinders with Raised Blue Graduations
Description
- Acc. to DIN 12681/ISO 6706 PMP
- Raised scale
- transparent
- Main point ring graduation
- Hexagonal base
- Conformity certified
Specifications
Specifications
| Material | PMP |
| Class | A |
| Disposable | Reusable |
| Graduations | 1 mL |
| Shape | Round |
| For Use With (Application) | Precise Liquid Measurement and Fluid Transfer in the Laboratory |
| Scale Color | Blue |
| Base Type | Hexagonal |
| Height (Metric) | 200 mm |
| Type | Graduated Measuring Cylinder |