Learn More
BINDER™ Class 3.1 Independent Temperature Safety Device
Product Code. 16464349 Shop All BINDER Products
Click to view available options
For Use With (Equipment):
BINDER Chambers FD 56/115/260/720
BINDER Chambers FED 56/115/260/720
Unit Size:
Each
Product Code. 16464349
Brand: BINDER™ 80121644
Only null left
Add to basket
Add to basket
Class 3.1 independent temperature safety device - with visual alarm (DIN 12880)
  • Class 3.1
  • With visual alarm
  • Safety devices also available in Class 2 and 3.3
Specifications
Description Class 3.1 independent temperature safety device - with visual alarm (DIN 12880)
For Use With (Equipment) BINDER Chambers FED 56/115/260/720
Material Plastic
Type Temperature Calibration Certificate
For Use With (Application) Temperature Regulation