Description
- Class 3.1
- With visual alarm
- Safety devices also available in Class 2 and 3.3
Specifications
Specifications
| Description | Class 3.1 independent temperature safety device - with visual alarm (DIN 12880) |
| For Use With (Equipment) | BINDER Chambers FD 56/115/260/720 |
| Material | Plastic |
| Type | Temperature Calibration Certificate |
| For Use With (Application) | Temperature Regulation |